siRNA Details
Gene Symbol: Thoc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285780.1 | NP_001272709.1 | 5 | 518 | ||
NM_025435.3 | NP_079711.1 | 4 | 440 | |||
GenBank mRNA | AK005176.1 | - | 4 | 367 | ||
AK007440.1 | - | 4 | 346 | |||
AK010958.1 | - | 4 | 381 | |||
AK013319.1 | - | 4 | 363 | |||
AK143106.1 | - | 2 | 1588 | |||
AK147003.1 | - | 5 | 421 | |||
BC027113.1 | - | 3 | 1388 | |||
BC042209.1 | - | 4 | 367 | |||
BC054419.1 | - | 4 | 400 | 4010343 | ||
BC116919.1 | - | 4 | 405 | 40126238 | ||
BC116945.1 | - | 4 | 405 | 40126264 | ||
CK329337.1 | - | 4 | 441 |

More Information
Other Species Targeted

Gene Ontology Categories:
