siRNA Details
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025437.4 | NP_079713.2 | 5, 6 | 543 | ||
GenBank mRNA | AK005185.1 | - | 5, 6 | 531 | ||
AK007301.1 | - | 5, 6 | 494 | |||
AK012233.1 | - | 5, 6 | 523 | |||
AK012714.1 | - | 5, 6 | 526 | |||
AK021283.1 | - | 5, 6 | 524 | |||
AK030774.1 | - | 547 | ||||
AK031113.1 | - | 555 | ||||
AK077710.1 | - | 5, 6 | 492 | |||
AK090053.1 | - | 509 | ||||
AK134827.1 | - | 468 | ||||
AK134969.1 | - | 468 | ||||
AK165574.1 | - | 509 | ||||
AK166629.1 | - | 5, 6 | 546 | |||
AK166852.1 | - | 5, 6 | 544 | |||
AK207164.1 | - | 4, 5 | 268 | |||
AK210743.1 | - | 4, 5 | 257 | |||
AK217305.1 | - | 4, 5 | 305 | |||
BC027284.1 | - | 5, 6 | 480 | 3156134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
