siRNA Details
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025447.4 | NP_079723.1 | 9 | 767 | ||
GenBank mRNA | AK007759.1 | - | 9 | 718 | ||
AK011544.1 | - | 9 | 724 | |||
AK015145.1 | - | 9 | 717 | |||
AK078186.1 | - | 7 | 1561 | |||
AK078401.1 | - | 7 | 1583 | |||
AK146188.1 | - | 9 | 738 | |||
AK161333.1 | - | 9 | 719 | |||
BC019799.1 | - | 9 | 740 | 5053500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
