siRNA Details
Gene Symbol: Camk2n1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025451.2 | NP_079727.1 | 1, 2 | 201 | ||
GenBank mRNA | AY523601.1 | - | 1, 2 | 163 |

More Information
Other Species Targeted

Gene Ontology Categories:
