siRNA Details
Gene Symbol: Ccdc28b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025455.2 | NP_079731.2 | 3 | 411 | ||
GenBank mRNA | AK003287.1 | - | 3 | 381 | ||
AK007427.1 | - | 2 | 188 | |||
AK028262.1 | - | 3 | 389 | |||
AK031278.1 | - | 1 | 137 | |||
AK158016.1 | - | 3 | 415 | |||
BC028873.1 | - | 3 | 403 | 4220355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
