siRNA Details
Gene Symbol: Fam134b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034851.2 | NP_001030023.1 | 4 | 517 | ||
NM_001277315.1 | NP_001264244.1 | 2 | 606 | |||
NM_001277316.1 | NP_001264245.1 | 2 | 189 | |||
NM_001277317.1 | NP_001264246.1 | 2 | 167 | |||
NM_001277318.1 | NP_001264247.1 | 2 | 139 | |||
NM_025459.3 | NP_079735.2 | 3 | 307 | |||
GenBank mRNA | AK007506.1 | - | 2 | 168 | ||
AK015182.1 | - | 3 | 129 | |||
AK146972.1 | - | 2 | 190 | |||
AK167327.1 | - | 2 | 607 | |||
AK195437.1 | - | 4 | 249 | |||
BC019494.1 | - | 3 | 246 | 4460992 | ||
BC054415.1 | - | 2 | 126 | 3487542 | ||
CB724233.1 | - | 4 | 636 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001034912, XM_006232083, XM_008760821, XM_006232084, BC089887.
Gene Ontology Categories:
