siRNA Details
Gene Symbol: Smim20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145433.1 | NP_001138905.1 | 2 | 284 | ||
GenBank mRNA | AK003139.1 | - | 2 | 219 | ||
AK007474.1 | - | 3 | 255 | |||
BC030079.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
