siRNA Details
Gene Symbol: Tmem218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025464.2 | NP_079740.1 | 3 | 218 | ||
GenBank mRNA | AK003010.1 | - | 3 | 215 | ||
AK007575.1 | - | 3 | 202 | |||
AK008902.1 | - | 3 | 189 | |||
AK161115.1 | - | 220 | ||||
BC034160.1 | - | 3 | 218 | 5049687 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
