siRNA Details
Gene Symbol: Tma16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025465.2 | NP_079741.1 | 2 | 152 | ||
GenBank mRNA | AK007637.1 | - | 2 | 152 | ||
AK009167.1 | - | 2 | 167 | |||
AK011532.1 | - | 2 | 163 | |||
AK031875.1 | - | 2 | 176 | |||
BC016246.1 | - | 2 | 142 | 4511374 | ||
BC034844.1 | - | 2 | 148 | 1496036 |

More Information
Other Species Targeted

Gene Ontology Categories:
