siRNA Details
Gene Symbol: Smim8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025471.2 | NP_079747.1 | 3 | 358 | ||
GenBank mRNA | AK005941.1 | - | 3 | 361 | ||
AK007303.1 | - | 2 | 116 | |||
AK007660.1 | - | 2 | 136 | |||
AK021236.1 | - | 2 | 153 | |||
AK079613.1 | - | 2 | 147 | |||
BC027508.1 | - | 2 | 106 | 1494969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
