siRNA Details
Gene Symbol: Haus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290808.1 | NP_001277737.1 | 1 | 153 | ||
NM_025475.3 | NP_079751.1 | 1 | 153 | |||
GenBank mRNA | AK010445.1 | - | 1 | 100 | ||
AK012086.1 | - | 1 | 152 | |||
AK012779.1 | - | 1 | 153 | |||
AK012960.1 | - | 1 | 100 | |||
AK132300.1 | - | 1 | 109 | |||
BC086677.1 | - | 1 | 91 | 30660268 | ||
BY711279.1 | - | 1 | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
