siRNA Details
Gene Symbol: Fam53c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175104.4 | NP_780313.1 | 4, 5 | 1314 | ||
GenBank mRNA | AK044469.1 | - | 4, 5 | 1118 | ||
AK135712.1 | - | 3, 4 | 1060 | |||
AK154214.1 | - | 4, 5 | 1118 | |||
AK166841.1 | - | 3, 4 | 1058 | |||
BC044896.1 | - | 3, 4 | 1039 | |||
BC057111.1 | - | 4, 5 | 1314 | 6417613 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008772055, XM_006254613, XM_001062862.
Gene Ontology Categories:
