siRNA Details
Gene Symbol: Tmem128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025480.3 | NP_079756.2 | 2 | 202 | ||
GenBank mRNA | AK012789.2 | - | 2 | 203 | ||
AK013227.1 | - | 2 | 202 | |||
AK076679.1 | - | 2 | 194 | |||
AK080853.1 | - | 2 | 203 | |||
AK162257.1 | - | 2 | 197 | |||
BC025197.1 | - | 2 | 361 | 5251991 | ||
BC060969.1 | - | 2 | 191 | 6704414 | ||
BC110310.1 | - | 2 | 185 | 5325405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
