siRNA Details
Gene Symbol: Cenpw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109747.1 | NP_001103217.1 | 2 | 239 | ||
GenBank mRNA | AK005988.1 | - | 2 | 221 | ||
AK011700.1 | - | 2 | 221 | |||
AK141272.1 | - | 2 | 221 | |||
BC050071.1 | - | 191 | ||||
BC147188.1 | - | 2 | 236 | 9056212 | ||
BC147189.1 | - | 2 | 236 | 9056237 | ||
BC147710.1 | - | 2 | 236 | 9087469 | ||
BC147721.1 | - | 2 | 236 | 9087480 | ||
BG804382.1 | - | 2 | 239 |

More Information
Other Species Targeted

Gene Ontology Categories:
