siRNA Details
Gene Symbol: Smurf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025481.2 | NP_079757.2 | 15 | 2200 | ||
GenBank mRNA | AK013082.1 | - | 2 | 247 | ||
AY685230.1 | - | 15 | 1880 | |||
BC023703.1 | - | 3 | 329 | |||
BC065796.1 | - | 1 | 44 | |||
BC138786.1 | - | 16 | 1765 | 8861808 | ||
BC138788.1 | - | 16 | 1776 | 8861810 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
