siRNA Details
Gene Symbol: 1700011A15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025487.3 | NP_079763.1 | 2 | 189 | ||
GenBank mRNA | AK005855.1 | - | 2 | 146 | ||
AK007046.1 | - | 2 | 161 | |||
BC048463.1 | - | 2 | 166 | 6742420 | ||
CN841812.1 | - | 2 | 193 |
