siRNA Details
Gene Symbol: Scp2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025490.2 | NP_079766.3 | 1 | 498 | ||
GenBank mRNA | AK005847.1 | - | 1 | 498 | ||
AK015138.1 | - | 1 | 449 | |||
BC048653.1 | - | 1 | 498 | 6773017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
