siRNA Details
Gene Symbol: Fam229b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183254.1 | NP_899077.1 | 5 | 535 | ||
GenBank mRNA | AK006353.1 | - | 5 | 535 | ||
AK006431.1 | - | 5 | 514 | |||
BC147171.1 | - | 4 | 464 | 9056241 | ||
BC147172.1 | - | 4 | 464 | 9056208 | ||
BC172004.1 | - | 4 | 461 | 9054290 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
