siRNA Details
Gene Symbol: Psenen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025498.2 | NP_079774.1 | 2, 3 | 274 | ||
GenBank mRNA | AK002440.1 | - | 2, 3 | 222 | ||
AK006284.1 | - | 2, 3 | 239 | |||
AK007621.1 | - | 2, 3 | 237 | |||
BC024347.1 | - | 2, 3 | 274 | 4984284 | ||
BC081443.1 | - | 2, 3 | 276 | 6814799 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
