siRNA Details
Gene Symbol: Eid3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025499.2 | NP_079775.2 | 1 | 629 | ||
GenBank mRNA | AK006438.1 | - | 1 | 598 | ||
AK007194.1 | - | 1 | 624 | |||
AK132745.1 | - | 1 | 594 | |||
AK161275.1 | - | 1 | 629 | |||
AK161307.1 | - | 1 | 629 | |||
BC099545.1 | - | 1 | 596 | 30917551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
