siRNA Details
Gene Symbol: Tmem177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175106.3 | NP_780315.1 | 2 | 605 | ||
GenBank mRNA | AK009043.1 | - | 2 | 610 | ||
AK076108.1 | - | 2 | 611 | |||
BC094344.1 | - | 2 | 605 | 6402386 | ||
BC131993.1 | - | 1 | 447 | 40130270 | ||
BC132021.1 | - | 1 | 447 | 40130298 |
