siRNA Details
Gene Symbol: Atp5sl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290487.1 | NP_001277416.1 | 2, 3 | 242 | ||
NM_025504.4 | NP_079780.2 | 2, 3 | 1167 | |||
GenBank mRNA | AK002370.1 | - | 2, 3 | 222 | ||
AK009150.1 | - | 2, 3 | 225 | |||
AK039669.1 | - | 2, 3 | 1168 | |||
BC024867.1 | - | 1, 2 | 198 | 4951552 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
