siRNA Details
Gene Symbol: Pla2g12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286948.1 | NP_001273877.1 | 2 | 342 | ||
NM_023196.4 | NP_075685.2 | 2 | 342 | |||
NM_183423.3 | NP_904359.1 | 2 | 125 | |||
GenBank mRNA | AK003183.1 | - | 2 | 302 | ||
AK009133.1 | - | 2 | 344 | |||
AK010011.1 | - | 2 | 332 | |||
AK010174.1 | - | 2 | 308 | |||
AK012414.1 | - | 2 | 270 | |||
AK163693.1 | - | 2 | 256 | |||
AK178670.1 | - | 2 | 50 | |||
AY007381.1 | - | 2 | 315 | |||
AY007382.1 | - | 2 | 125 | |||
BC026812.1 | - | 2 | 307 | 4456431 | ||
BC051117.1 | - | 2 | 342 | 6306803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
