siRNA Details
Gene Symbol: Riiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025506.2 | NP_079782.1 | 5, 6 | 385 | ||
GenBank mRNA | AK009192.1 | - | 5, 6 | 387 | ||
AK019569.1 | - | 3, 4 | 195 | |||
BC107021.1 | - | 4, 5 | 772 | 40049310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
