siRNA Details
Gene Symbol: Knop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168218.1 | NP_001161690.1 | 1, 2 | 379 | ||
NM_001168219.1 | NP_001161691.1 | 1, 2 | 379 | |||
NM_023197.3 | NP_075686.2 | 1, 2 | 379 | |||
GenBank mRNA | AF034580.1 | - | 2, 3 | 546 | ||
AK009230.1 | - | 1, 2 | 40 | |||
AK083845.1 | - | 1, 2 | 66 | |||
AK166956.1 | - | 1, 2 | 65 |
