siRNA Details
Gene Symbol: Knop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168218.1 | NP_001161690.1 | 2 | 492 | ||
NM_001168219.1 | NP_001161691.1 | 2 | 492 | |||
NM_001168220.1 | NP_001161692.1 | 2 | 342 | |||
NM_023197.3 | NP_075686.2 | 2 | 492 | |||
GenBank mRNA | AF034580.1 | - | 3 | 659 | ||
AK009230.1 | - | 2 | 153 | |||
AK083845.1 | - | 2 | 179 | |||
AK166956.1 | - | 2 | 178 | |||
BC055941.1 | - | 2 | 332 | 4009257 |
