siRNA Details
Gene Symbol: Ostc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025509.3 | NP_079785.1 | 2, 3 | 308 | ||
GenBank mRNA | AK011085.1 | - | 2, 3 | 260 | ||
AK013011.1 | - | 2, 3 | 267 | |||
AK135332.1 | - | 2, 3 | 264 | |||
BC014799.1 | - | 2, 3 | 283 | |||
BC021935.1 | - | 2, 3 | 282 | 5149713 | ||
BY228396.1 | - | 2, 3 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
