siRNA Details
Gene Symbol: Ostc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025509.3 | NP_079785.1 | 1, 2 | 211 | ||
GenBank mRNA | AK009237.1 | - | 1, 2 | 186 | ||
AK011085.1 | - | 1, 2 | 163 | |||
AK013011.1 | - | 1, 2 | 170 | |||
AK135332.1 | - | 1, 2 | 167 | |||
BC014799.1 | - | 1, 2 | 184 | |||
BC021935.1 | - | 1, 2 | 185 | 5149713 | ||
BY228396.1 | - | 1, 2 | 212 |

More Information
Other Species Targeted

Gene Ontology Categories:
