siRNA Details
Gene Symbol: Zfand1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025512.2 | NP_079788.2 | 7 | 602 | ||
GenBank mRNA | AK009240.1 | - | 7 | 616 | ||
AK017696.1 | - | 7 | 612 | |||
AK030472.1 | - | 7 | 610 | |||
AK077873.1 | - | 7 | 612 | |||
AK081710.1 | - | 1 | 3696 | |||
BC023352.1 | - | 7 | 594 | |||
BC025216.1 | - | 7 | 594 | |||
BC046585.1 | - | 7 | 602 | 6314252 | ||
BC089494.1 | - | 7 | 614 | 30504018 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
