siRNA Details
Gene Symbol: 2310011J03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025521.3 | NP_079797.1 | 2 | 1195 | ||
GenBank mRNA | AK046452.1 | - | 2 | 1156 | ||
AK140230.1 | - | 2 | 1156 | |||
AK180888.1 | - | 1 | 417 | |||
BC023429.1 | - | 2 | 1162 | 5042442 | ||
BY596174.1 | - | 1 | 27 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
