siRNA Details
Gene Symbol: Ndufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025523.1 | NP_079799.1 | 4 | 330 | ||
GenBank mRNA | AK003365.1 | - | 4 | 316 | ||
AK009393.1 | - | 4 | 330 | |||
AK018705.1 | - | 4 | 314 | |||
AK028011.1 | - | 314 | ||||
BC051933.1 | - | 4 | 311 | |||
BC092141.1 | - | 4 | 322 | 6814605 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
