siRNA Details
Gene Symbol: Cox14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183256.3 | NP_899079.1 | 2 | 296 | ||
GenBank mRNA | AK002816.1 | - | 2 | 159 | ||
AK004378.1 | - | 2 | 115 | |||
AK009401.1 | - | 2 | 268 | |||
BC059905.1 | - | 2 | 248 | 6823331 | ||
BC092142.1 | - | 2 | 197 | 6814201 | ||
BP774190.1 | - | 2 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
