siRNA Details
Gene Symbol: Srp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025527.3 | NP_079803.2 | 3 | 226 | ||
GenBank mRNA | AK004118.1 | - | 3 | 228 | ||
AK009412.1 | - | 3 | 190 | |||
AK010760.1 | - | 3 | 225 | |||
AK037009.1 | - | 2 | 2696 | |||
AK135508.1 | - | 4 | 353 | |||
AK152322.1 | - | 3 | 205 | |||
AK162107.1 | - | 3 | 196 | |||
BC051932.1 | - | 3 | 193 | 5710637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
