siRNA Details
Gene Symbol: Nudt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025529.3 | NP_079805.1 | 2 | 301 | ||
GenBank mRNA | AK002605.1 | - | 2 | 239 | ||
AK009700.1 | - | 2 | 239 | |||
AK045197.1 | - | 2 | 302 | |||
AK170076.1 | - | 2 | 241 | |||
BC056443.1 | - | 2 | 241 | 6822891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
