siRNA Details
Gene Symbol: Slmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025531.2 | NP_079807.1 | 2, 3 | 284 | ||
GenBank mRNA | AK003043.1 | - | 2, 3 | 258 | ||
AK008542.1 | - | 2, 3 | 244 | |||
AK013197.1 | - | 2, 3 | 251 | |||
AK150668.1 | - | 2, 3 | 260 | |||
BC004775.1 | - | 1, 2 | 82 | |||
BC026968.1 | - | 2, 3 | 284 | 4235395 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001009543, BC060590.
Gene Ontology Categories:
