siRNA Details
Gene Symbol: Sln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025540.2 | NP_079816.1 | 2 | 494 | ||
GenBank mRNA | AK008863.1 | - | 2 | 497 | ||
AK008896.1 | - | 2 | 495 | |||
AK009005.1 | - | 2 | 497 | |||
AK009809.1 | - | 582 | ||||
BC028496.1 | - | 2 | 475 | 1397989 | ||
CA460968.1 | - | 2 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
