siRNA Details
Gene Symbol: Mrpl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025553.4 | NP_079829.1 | 2 | 244 | ||
GenBank mRNA | AB049639.1 | - | 2 | 222 | ||
AK002797.1 | - | 2 | 199 | |||
AK010329.1 | - | 2 | 236 | |||
AK167247.1 | - | 2 | 212 | |||
BC046750.1 | - | 2 | 197 | 5692108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
