siRNA Details
Gene Symbol: Cyb5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025558.5 | NP_079834.2 | 2, 3 | 342 | ||
GenBank mRNA | AK003333.2 | - | 2, 3 | 343 | ||
AK003815.1 | - | 2, 3 | 332 | |||
AK007780.1 | - | 362 | ||||
AK019263.1 | - | 2, 3 | 342 | |||
AK052810.1 | - | 2, 3 | 343 | |||
AK083359.1 | - | 2, 3 | 188 | |||
AK088970.1 | - | 2, 3 | 344 | |||
AK166544.1 | - | 2, 3 | 320 | |||
AK168422.1 | - | 2, 3 | 332 | |||
BC046479.1 | - | 2, 3 | 305 | |||
BC046810.1 | - | 2, 3 | 308 | |||
BC048147.1 | - | 2, 3 | 332 | |||
BC054749.1 | - | 2, 3 | 363 | 6410090 | ||
BC058812.1 | - | 2, 3 | 305 | 6530835 | ||
BC062980.1 | - | 2, 3 | 354 | 6842939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
