siRNA Details
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190322.2 | NP_001177251.1 | 4 | 438 | ||
NM_001190323.2 | NP_001177252.1 | 4 | 314 | |||
NM_001190324.2 | NP_001177253.1 | 4 | 269 | |||
NM_001285804.1 | NP_001272733.1 | 4 | 469 | |||
NM_181391.4 | NP_852056.1 | 4 | 483 | |||
GenBank mRNA | AA980189.1 | - | 4 | 266 | ||
AK003308.1 | - | 4 | 315 | |||
AK007844.1 | - | 4 | 458 | |||
AK013200.1 | - | 4 | 456 | |||
AK145105.1 | - | 4 | 458 | |||
AK168647.1 | - | 4 | 413 | |||
AK169018.1 | - | 4 | 458 | |||
AV042014.2 | - | 2 | 78 | |||
BC030366.1 | - | 4 | 441 | 5370748 | ||
BX527959.1 | - | 4 | 230 | |||
BY578448.1 | - | 3 | 201 | |||
CB057699.1 | - | 4 | 395 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
