siRNA Details
Gene Symbol: Cdc26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139291.3 | NP_647452.1 | 4 | 458 | ||
GenBank mRNA | AK004518.1 | - | 1 | 98 | ||
AK012627.1 | - | 4 | 459 | |||
AK047055.1 | - | 478 | ||||
AK086692.1 | - | 937 | ||||
AK135556.1 | - | 458 | ||||
BC005775.1 | - | 3 | 385 | 3602527 | ||
BC114350.2 | - | 2 | 245 | 40053418 |

More Information
Other Species Targeted

Gene Ontology Categories:
