siRNA Details
Gene Symbol: Pam16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025571.1 | NP_079847.1 | 4 | 373 | ||
GenBank mRNA | AB073618.1 | - | 4 | 346 | ||
AF349454.1 | - | 4 | 358 | |||
AK003227.1 | - | 4 | 363 | |||
AK008380.1 | - | 4 | 373 | |||
BC024346.1 | - | 4 | 353 | 4989038 | ||
BC096419.1 | - | 4 | 362 | 1546951 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
