siRNA Details
Gene Symbol: Pyurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025574.3 | NP_079850.1 | 2 | 293 | ||
GenBank mRNA | AK003713.1 | - | 2 | 294 | ||
AK008978.1 | - | 2 | 293 | |||
AK011515.1 | - | 2 | 230 | |||
AK029193.1 | - | 269 | ||||
AK032381.1 | - | 2 | 286 | |||
BC029232.1 | - | 2 | 257 | 4164517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
