siRNA Details
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253757.1 | NP_001240686.1 | 4 | 733 | ||
NM_023210.4 | NP_075699.3 | 4 | 733 | |||
GenBank mRNA | AB037685.1 | - | 4 | 408 | ||
AK012759.1 | - | 4 | 700 | |||
AK049647.1 | - | 4 | 700 | |||
AK076049.1 | - | 4 | 560 | |||
AK088401.1 | - | 4 | 713 | |||
AK147888.1 | - | 4 | 699 | |||
AK159446.1 | - | 4 | 700 | |||
AK168380.1 | - | 4 | 682 | |||
AK172146.1 | - | 4 | 747 | |||
AK193570.1 | - | 2 | 195 | |||
AK195817.1 | - | 2 | 104 | |||
AK199406.1 | - | 2 | 195 | |||
AK202251.1 | - | 2 | 197 | |||
AK204175.1 | - | 2 | 195 | |||
BC005690.1 | - | 4 | 668 | 3596597 | ||
BC080684.1 | - | 4 | 673 | 6493476 | ||
CF952181.1 | - | 4 | 404 | |||
U89345.2 | - | 4 | 694 |

More Information
Other Species Targeted

Gene Ontology Categories:
