siRNA Details
Gene Symbol: Ctrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025583.2 | NP_079859.2 | 5 | 364 | ||
GenBank mRNA | AK003060.1 | - | 5 | 373 | ||
AK003079.1 | - | 5 | 365 | |||
AK007566.1 | - | 5 | 364 | |||
AK007765.1 | - | 5 | 364 | |||
AK007815.1 | - | 5 | 365 | |||
AK008644.2 | - | 5 | 365 | |||
AK008729.1 | - | 5 | 365 | |||
AK008888.1 | - | 5 | 365 | |||
AK008927.1 | - | 5 | 365 | |||
AK018554.1 | - | 5 | 366 | |||
BC061083.1 | - | 5 | 364 | 30316047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
