siRNA Details
Gene Symbol: Rps21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025587.2 | NP_079863.1 | 5 | 285 | ||
GenBank mRNA | AA049954.1 | - | 4 | 211 | ||
AK007846.1 | - | 5 | 248 | |||
AK007850.1 | - | 5 | 240 | |||
AK010610.1 | - | 5 | 286 | |||
AK010637.1 | - | 5 | 253 | |||
AK012488.1 | - | 5 | 248 | |||
BC027563.1 | - | 5 | 233 | 3468174 | ||
BC086912.1 | - | 5 | 250 | 6758965 | ||
BU534365.1 | - | 5 | 288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
