siRNA Details
Gene Symbol: Fam136a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025591.2 | NP_079867.1 | 3 | 307 | ||
GenBank mRNA | AK005280.1 | - | 3 | 267 | ||
AK008550.1 | - | 3 | 273 | |||
AK141279.1 | - | 3 | 267 | |||
BC013621.1 | - | 3 | 255 | |||
BC024049.1 | - | 3 | 307 | 5100000 | ||
BC103775.1 | - | 3 | 308 | 6485116 | ||
CT010180.1 | - | 3 | 257 |

More Information
Other Species Targeted

Gene Ontology Categories:
