siRNA Details
Gene Symbol: Rpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025592.3 | NP_079868.1 | 2, 3 | 181 | ||
GenBank mRNA | AK010639.2 | - | 2, 3 | 182 | ||
AK012335.1 | - | 2, 3 | 182 | |||
AK151707.1 | - | 2, 3 | 176 | |||
AK169650.1 | - | 2, 3 | 182 | |||
BC025884.1 | - | 2, 3 | 163 | |||
BC094634.1 | - | 2, 3 | 165 | 1333082 | ||
BC125652.1 | - | 2, 3 | 163 | 40130167 | ||
BC125656.1 | - | 2, 3 | 163 | 40130171 |

More Information
Other Species Targeted

Gene Ontology Categories:
