siRNA Details
Gene Symbol: Ndufb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025597.2 | NP_079873.1 | 2 | 151 | ||
GenBank mRNA | AK007930.1 | - | 2 | 150 | ||
AK009008.1 | - | 2 | 113 | |||
AK012320.1 | - | 2 | 152 | |||
AK013608.1 | - | 2 | 137 | |||
AK138123.1 | - | 2 | 149 | |||
BC028669.1 | - | 2 | 307 | |||
BC036989.1 | - | 2 | 148 | 4989233 |

More Information
Other Species Targeted

Gene Ontology Categories:
