siRNA Details
Gene Symbol: Chtop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293776.1 | NP_001280705.1 | 6 | 997 | ||
NM_001293777.1 | NP_001280706.1 | 6 | 994 | |||
NM_001293778.1 | NP_001280707.1 | 5 | 859 | |||
NM_001293779.1 | NP_001280708.1 | 5 | 856 | |||
NM_001293780.1 | NP_001280709.1 | 4 | 2142 | |||
NM_001293781.1 | NP_001280710.1 | 4 | 2139 | |||
NM_001293782.1 | NP_001280711.1 | 6 | 1994 | |||
NM_023215.6 | NP_075704.2 | 5 | 2280 | |||
GenBank mRNA | AB041654.1 | - | 4 | 2094 | ||
AK004558.2 | - | 5 | 2226 | |||
AK005377.1 | - | 5 | 821 | |||
AK008885.1 | - | 3 | 293 | |||
AK010866.1 | - | 6 | 979 | |||
AK047914.1 | - | 6 | 971 | |||
AK077960.1 | - | 4 | 2128 | |||
AK132769.1 | - | 6 | 970 | |||
AK135914.1 | - | 5 | 835 | |||
AK149010.1 | - | 3 | 468 | |||
AK152492.1 | - | 6 | 950 | |||
AK211997.1 | - | 3 | 317 | |||
BC004582.1 | - | 1 | 129 | |||
BC051939.1 | - | 6 | 1948 | |||
BC066837.1 | - | 5 | 2233 | |||
BC083333.1 | - | 4 | 583 | |||
BC108396.1 | - | 5 | 626 | |||
BC120651.1 | - | 4 | 1517 | 40129574 | ||
BC144775.1 | - | 3 | 1379 | 9053323 |

More Information
Other Species Targeted

Gene Ontology Categories:
