siRNA Details
Gene Symbol: Pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023217.4 | NP_075706.1 | 5 | 1007 | ||
GenBank mRNA | AJ278829.1 | - | 5 | 939 | ||
AK012658.1 | - | 5 | 939 | |||
AK047835.1 | - | 5 | 951 | |||
AK049632.1 | - | 5 | 930 | |||
AK208078.1 | - | 1 | 245 | |||
BC043041.1 | - | 5 | 917 | |||
BC051938.1 | - | 5 | 936 | 5717365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
